Learn More
CPC Scientific H-Ser-Pro-Lys-Thr-Met-Arg-Asp-Ser-Gly-Cys-Phe-Gly-Arg-Arg-Leu-Asp-Arg-Ile-Gly-Ser-Leu-Ser-Gly-Leu-Gly-Cys-Asn-Val-Leu-Arg-Arg-Tyr-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NATR015B
Encompass
Brain natriuretic peptide or B-type natriuretic peptide (BNP) (also ventricular natriuretic peptide) is a 32-amino acid peptide secreted by heart ventricles in response to excessive stretching of heart muscle cells (cardiomyocytes).ONE-LETTER SEQUENCE: SPKTMRDSGCFGRRLDRIGSLSGLGCNVLRRY (Cys10 and 26 bridge)MOLECULAR FORMULA: C149H250N52O44S3MOLECULAR WEIGHT:3570.2STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [117345-87-6], RESEARCH AREA: CardiovascularREFERENCES: T. Sudoh et al., BBRC, 155, 726 (1988)
Catalog No. 50-210-9869