Learn More
CPC Scientific H-Ser-Gln-Asp-Ser-Ala-Phe-Arg-Ile-Gln-Glu-Arg-Leu-Arg-Asn-Ser-Lys-Met-Ala-His-Ser-Ser-Ser-Cys-Phe-Gly-Gln-Lys-Ile-Asp-Arg-Ile-Gly-Ala-Val-Ser-Arg-Leu-Gly-Cys-Asp-Gly-Leu-Arg-Leu-Phe-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502109867
Encompass
ONE-LETTER SEQUENCE: SQDSAFRIQERLRNSKMAHSSSCFGQKIDRIGAVSRLGCDGLRLF (Cys23 and 39 bridge)MOLECULAR FORMULA: C213H349N71O65S3MOLECULAR WEIGHT:5040.8STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [123337-89-3], SYNONYMS: BNP-45 (rat), Brain Natriuretic Peptide-45 (rat)RESEARCH AREA: CardiovascularREFERENCES: M. Aburaya et al., BBRC, 163, 226 (1989)
Catalog No. 50-210-9867 Supplier CPC Scientific Supplier No. NATR014B
ONE-LETTER SEQUENCE: SQDSAFRIQERLRNSKMAHSSSCFGQKIDRIGAVSRLGCDGLRLF (Cys23 and 39 bridge)MOLECULAR FORMULA: C213H349N71O65S3MOLECULAR WEIGHT:5040.8STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [123337-89-3], SYNONYMS: BNP-45 (rat), Brain Natriuretic Peptide-45 (rat)RESEARCH AREA: CardiovascularREFERENCES: M. Aburaya et al., BBRC, 163, 226 (1989)