Learn More
TARGETMOL CHEMICALS INC BNTA 5MG
SDP
Catalog No. 502579970
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BNTA is a potent extracellular matrix (ECM) modulator. In a rat model of arthritis BNTA modulates arthritis by promoting the synthesis of cartilage structural molecules on chondrocytes through the induction of superoxide dismutase 3 (SOD3) and regulating chondrogenesis through superoxide anion elimination. purity: 99%
Catalog No. 50-257-9970 Supplier TARGETMOL CHEMICALS INC Supplier No. T407955MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BNTA is a potent extracellular matrix (ECM) modulator. In a rat model of arthritis BNTA modulates arthritis by promoting the synthesis of cartilage structural molecules on chondrocytes through the induction of superoxide dismutase 3 (SOD3) and regulating chondrogenesis through superoxide anion elimination. purity: 99%