Learn More
Medchemexpress LLC HY-136651 5mg Medchemexpress, BNTA CAS:685119-25-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1366515MG
Encompass_Preferred
Medchemexpress, HY-136651 5mg BNTA CAS:685119-25-9 BNTA, a potent extracellular matrix (ECM) modulator, facilitates cartilage structural molecule synthesis on chondrocytes by activating superoxide dismutase 3 (SOD3). BNTA shows a promising potential for osteoarthritis alleviation by modulating cartilage generation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1452