Learn More
Medchemexpress LLC HY-135960 10mg Medchemexpress, BO-264 CAS:2408648-20-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13596010MG
Encompass_Preferred
Medchemexpress, HY-135960 10mg BO-264 CAS:2408648-20-2 BO-264 is a highly potent and orally active transforming acidic coiled-coil 3 (TACC3) inhibitor with an IC50 of 188 nM and a Kd of 1.5 nM. BO-264 specifically blocks the function of FGFR3-TACC3 fusion protein. BO-264 induces spindle assembly checkpoint (SAC)-dependent mitotic arrest, DNA damage and apoptosis. BO-264 has broad-spectrum antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0388