Learn More
World Precision Instrument Board Frame Clamp (pkg of 4)
SDP
Catalog No. 502539536
Encompass
Non-Magnetic Bases, Stainless Steel Rods, & Clamps
  • Opens to 8.5mm
  • Combine rods to build lab frame
Catalog No. 50-253-9536 Supplier World Precision Instrument Supplier No. 5030824
Non-Magnetic Bases, Stainless Steel Rods, & Clamps
  • Opens to 8.5mm
  • Combine rods to build lab frame