Learn More
Enterprise Technology Solutions MAGNETIC GLASS DRY ERASE BOARD VALUE PACK, 72 X 36, WHITE
SDP
Catalog No. 502051779
Encompass_Preferred
White frosted magnetic glass dry erase board is a perfect and functional solution for the frequent and heavy usage areas of your office, school classroom, home or any commercial space. Clean, contemporary and frameless product design lines will add a touch of subtle style to your wall. The modern frameless feature enables you to hang multiple glass boards adjacent to each other and expand the writing surface to fit your custom space needs. The board's premium magnetic ultimate glass writing surface will never stain or ghost. All ink erases easily with a simple eraser and the glass surface continuously outperforms all traditional melamine or painted steel dry erase writing surfaces. Occasionally, use any dry erase board spray or wet dry erase board wipes after continuous use to clean the writing surface. The glass magnetic surface works with high energy rare earth magnets.
Catalog No. 50-205-1779 Supplier Enterprise Technology Solutions Supplier No. UBR3973U0001
White frosted magnetic glass dry erase board is a perfect and functional solution for the frequent and heavy usage areas of your office, school classroom, home or any commercial space. Clean, contemporary and frameless product design lines will add a touch of subtle style to your wall. The modern frameless feature enables you to hang multiple glass boards adjacent to each other and expand the writing surface to fit your custom space needs. The board's premium magnetic ultimate glass writing surface will never stain or ghost. All ink erases easily with a simple eraser and the glass surface continuously outperforms all traditional melamine or painted steel dry erase writing surfaces. Occasionally, use any dry erase board spray or wet dry erase board wipes after continuous use to clean the writing surface. The glass magnetic surface works with high energy rare earth magnets.