Learn More
Med Vet International BOAS Surgery Kits - BOAS Surgery Mouth Gag Complete Kit, J1708, Each
Catalog No. 5000585296
Encompass_Preferred
"For Brachycephalic obstructive airway syndrome. There has been a rapid increase of Brachycephalic breeds such as French bulldogs. This has resulted in a larger number of surgeries to correct conditions associated with these breeds. The J1708 BOAS Surgery Mouth Gag Complete Kit. For Brachycephalic obstructive airway syndrome. There has been a rapid increase of Brachycephalic breeds such as French bulldogs. This has resulted in a larger number of surgeries to correct conditions associated with these breeds. This mouth gag system greatly improves visualization and surgical access. Special Design Features: • Fully adjustable tilting base to support the upper body of the patient at the desired angle to maximize visualization and access • Tilting base supported by sturdy support rods for excellent stability and optimal weight capacity • Two adjustable horizontal bars to retain the patient's open mouth in desired position
Catalog No. 50-005-85296 Supplier Med Vet International Supplier No. JOR1708
"For Brachycephalic obstructive airway syndrome. There has been a rapid increase of Brachycephalic breeds such as French bulldogs. This has resulted in a larger number of surgeries to correct conditions associated with these breeds. The J1708 BOAS Surgery Mouth Gag Complete Kit. For Brachycephalic obstructive airway syndrome. There has been a rapid increase of Brachycephalic breeds such as French bulldogs. This has resulted in a larger number of surgeries to correct conditions associated with these breeds. This mouth gag system greatly improves visualization and surgical access. Special Design Features: • Fully adjustable tilting base to support the upper body of the patient at the desired angle to maximize visualization and access • Tilting base supported by sturdy support rods for excellent stability and optimal weight capacity • Two adjustable horizontal bars to retain the patient's open mouth in desired position