Learn More
Cayman Chemical BOB1OBF1CTerm RabbIt M 100uL
SDP
Catalog No. 502948269
Encompass_Preferred
A GRK inhibitor (IC50s 3.9 0.15 and 0.38 M for GRK1 GRK2 and GRK5 respectively) selective for GRKs over PKA (IC50 120 M) increases contractility in isoproterenol-stimulated isolated mouse cardiomyocytes at 500 nM prevents the desensitization of histamine H1 receptor- and purinergic P2Y2 receptor-driven phospholipase C signaling in human ULTR myometrial cells and isolated rat MSMCs respectively (IC50s 3.09 and 2.95 M respectively)
Catalog No. 50-294-8269 Supplier Cayman Chemical Supplier No. 32303100 UL
A GRK inhibitor (IC50s 3.9 0.15 and 0.38 M for GRK1 GRK2 and GRK5 respectively) selective for GRKs over PKA (IC50 120 M) increases contractility in isoproterenol-stimulated isolated mouse cardiomyocytes at 500 nM prevents the desensitization of histamine H1 receptor- and purinergic P2Y2 receptor-driven phospholipase C signaling in human ULTR myometrial cells and isolated rat MSMCs respectively (IC50s 3.09 and 2.95 M respectively)