Learn More
Medchemexpress LLC Bobcat339 | 2280037-51-4 | 99.4% | 297.74 g·mol⁻¹ | C16H12ClN3O | 5 MG
SDP
Catalog No. 5000263397
Encompass_Preferred
Bobcat339 is a cytosine-based small-molecule inhibitor of TET enzymes used in epigenetic research. It inhibits TET1 and TET2 with reported IC50 values of 33 μM and 73 μM, and is provided in milligram quantities for laboratory studies.
  • Cytosine-based inhibitor of TET enzymes.
  • Reported IC50 values: 33 μM for TET1 and 73 μM for TET2.
  • High purity (99.44%) suitable for research applications.
  • Available in multiple milligram-scale packaging for experimental flexibility.
  • Useful for studying DNA demethylation and epigenetic regulation.
  • Datasheet available with chemical, physical, and handling information.
Catalog No. 50-002-63397 Supplier Medchemexpress LLC Supplier No. HY1115585MG
Bobcat339 is a cytosine-based small-molecule inhibitor of TET enzymes used in epigenetic research. It inhibits TET1 and TET2 with reported IC50 values of 33 μM and 73 μM, and is provided in milligram quantities for laboratory studies.
  • Cytosine-based inhibitor of TET enzymes.
  • Reported IC50 values: 33 μM for TET1 and 73 μM for TET2.
  • High purity (99.44%) suitable for research applications.
  • Available in multiple milligram-scale packaging for experimental flexibility.
  • Useful for studying DNA demethylation and epigenetic regulation.
  • Datasheet available with chemical, physical, and handling information.