Learn More
Medchemexpress LLC HY-111558A 10mg Medchemexpress, Bobcat339 hydrochloride CAS: Purity:>98%
SDP
Catalog No. 501968920
Encompass_Preferred
Medchemexpress, HY-111558A 10mg Bobcat339 HCl CAS: Bobcat339 hydrochloride is a cytosine-based TET enzyme inhibitor with IC50 of 33 μM (TET1) and 73 μM (TET2). It is useful to the field of epigenetics and serves as a starting point for new therapeutics that target DNA methylation and gene transcription. The TET enzymes is responsible for catalyzing the reverse process that is DNA demethylation by recognizing 5-methylcytosine and oxidizing the methyl group via an Fe(II)/alpha-ketoglutarate-dependent mechanism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8920 Supplier Medchemexpress LLC Supplier No. HY111558A10MG
Medchemexpress, HY-111558A 10mg Bobcat339 HCl CAS: Bobcat339 hydrochloride is a cytosine-based TET enzyme inhibitor with IC50 of 33 μM (TET1) and 73 μM (TET2). It is useful to the field of epigenetics and serves as a starting point for new therapeutics that target DNA methylation and gene transcription. The TET enzymes is responsible for catalyzing the reverse process that is DNA demethylation by recognizing 5-methylcytosine and oxidizing the methyl group via an Fe(II)/alpha-ketoglutarate-dependent mechanism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.