Learn More
Medchemexpress LLC Bobcat339 hydrochloride | 2436747-44-1 | 99.6% | 334.20 | 25 MG
SDP
Catalog No. 5000439531
Encompass_Preferred
Bobcat339 hydrochloride is a potent and selective cytosine-based inhibitor of the TET enzyme. It is considered useful in the field of epigenetics and can serve as a starting point for new therapeutics targeting DNA methylation and gene transcription.
  • Potent and selective inhibitor of TET enzyme
  • IC50s of 33 μM for TET1 and 73 μM for TET2
  • Significantly reduces global 5hmC levels in HT-22 cells
  • Useful for epigenetics research
  • Serves as a starting point for new therapeutics targeting DNA methylation and gene transcription
Catalog No. 50-004-39531 Supplier Medchemexpress LLC Supplier No. 25MGHY111558A
Bobcat339 hydrochloride is a potent and selective cytosine-based inhibitor of the TET enzyme. It is considered useful in the field of epigenetics and can serve as a starting point for new therapeutics targeting DNA methylation and gene transcription.
  • Potent and selective inhibitor of TET enzyme
  • IC50s of 33 μM for TET1 and 73 μM for TET2
  • Significantly reduces global 5hmC levels in HT-22 cells
  • Useful for epigenetics research
  • Serves as a starting point for new therapeutics targeting DNA methylation and gene transcription