Learn More
Medchemexpress LLC Boc-12-Ado-OH 25g
SDP
Catalog No. 5000206809
Encompass_Preferred
Boc-12-Ado-OH can be used as a PROTAC linker in the synthesis of PROTACs Boc-12-Ado-OH is an alkane chain with terminal carboxlic acid and Boc-protected amino groups The terminal carboxylic acid can react with primary amine groups in the presence of activators (e g EDC or HATU) to form a stable amide bond The Boc group can be deprotected under mild acidic conditions to form the free amine
Catalog No. 50-002-06809 Supplier Medchemexpress LLC Supplier No. 25GHYW101723
Boc-12-Ado-OH can be used as a PROTAC linker in the synthesis of PROTACs Boc-12-Ado-OH is an alkane chain with terminal carboxlic acid and Boc-protected amino groups The terminal carboxylic acid can react with primary amine groups in the presence of activators (e g EDC or HATU) to form a stable amide bond The Boc group can be deprotected under mild acidic conditions to form the free amine