Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BOC-(1R,3S)-3-aminocyclopentane carboxylic acid | 161660-94-2 | 229.28 | 100 MG
SDP

Catalog No. 5000448542
Encompass_Preferred

BOC-(1R,3S)-3-aminocyclopentane carboxylic acid is a synthetic intermediate useful for pharmaceutical synthesis. It is for research use only and not intended for sale to patients.

  • Useful for pharmaceutical synthesis
  • For research use only
  • Purity of 98.0%
  • Molecular weight is 229.28
  • White to off-white solid
  • Store at room temperature for 3 years
  • In solvent, store at -80°C for 2 years or -20°C for 1 year
  • Soluble in DMSO

Catalog No. 50-004-48542 Supplier Medchemexpress LLC Supplier No. HYW004812100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BOC-(1R,3S)-3-aminocyclopentane carboxylic acid is a synthetic intermediate useful for pharmaceutical synthesis. It is for research use only and not intended for sale to patients.

  • Useful for pharmaceutical synthesis
  • For research use only
  • Purity of 98.0%
  • Molecular weight is 229.28
  • White to off-white solid
  • Store at room temperature for 3 years
  • In solvent, store at -80°C for 2 years or -20°C for 1 year
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.