Learn More
Medchemexpress LLC BOC-(1S,3R)-3-aminocyclopentanecarboxylic acid | 261165-05-3 | 229.27 | 1 G
SDP
Catalog No. 5000446219
Encompass_Preferred
BOC-(1S,3R)-3-aminocyclopentanecarboxylic acid, also known as (1S,3R)-3-[(tert-Butoxycarbonyl)amino]cyclopentanecarboxylic acid, is a synthetic intermediate widely used in pharmaceutical synthesis.
  • Used as a synthetic intermediate for pharmaceutical synthesis.
  • Appears as a white to off-white solid.
  • Stable for 3 years when stored at room temperature.
  • Stable for 2 years at -80°C or 1 year at -20°C when stored in solvent.
Catalog No. 50-004-46219 Supplier Medchemexpress LLC Supplier No. HYW0062021G
BOC-(1S,3R)-3-aminocyclopentanecarboxylic acid, also known as (1S,3R)-3-[(tert-Butoxycarbonyl)amino]cyclopentanecarboxylic acid, is a synthetic intermediate widely used in pharmaceutical synthesis.
  • Used as a synthetic intermediate for pharmaceutical synthesis.
  • Appears as a white to off-white solid.
  • Stable for 3 years when stored at room temperature.
  • Stable for 2 years at -80°C or 1 year at -20°C when stored in solvent.