Learn More
Medchemexpress LLC Boc-6-aminohexanoic acid | 6404-29-1 | 231.29 | 100 G
SDP
Catalog No. 5000446420
Encompass_Preferred
Boc-6-aminohexanoic acid is an alkyl/ether-based PROTAC linker utilized in the synthesis of PROTACs. These compounds employ two distinct ligands connected by a linker, targeting an E3 ubiquitin ligase and a specific protein to facilitate its degradation via the intracellular ubiquitin-proteasome system.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs exploit the ubiquitin-proteasome system to degrade target proteins
Catalog No. 50-004-46420 Supplier Medchemexpress LLC Supplier No. HYW007529100G
Boc-6-aminohexanoic acid is an alkyl/ether-based PROTAC linker utilized in the synthesis of PROTACs. These compounds employ two distinct ligands connected by a linker, targeting an E3 ubiquitin ligase and a specific protein to facilitate its degradation via the intracellular ubiquitin-proteasome system.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs exploit the ubiquitin-proteasome system to degrade target proteins