Learn More
Bachem Boc-Ala-Ala-pNA
SDP
Supplier:  Bachem 4006301.1000
Encompass_Preferred
Boc-Ala-Ala-pNA, 50439-35-5, C17H24N4O6, Mr 380.40, 1g. Boc-AA-pNA is cleaved by serine proteases.
Catalog No. 50-194-7471