Learn More
Medchemexpress LLC Boc-AEVD-CHO | 220094-15-5 | 98.3% | 516.54 | 10 MG
SDP
Catalog No. 5000399163
Encompass_Preferred
Boc-AEVD-CHO is a Caspase 8 inhibitor useful in the study of apoptosis and immune and inflammatory diseases.
  • Inhibits Caspase 8
  • Useful in apoptosis studies
  • Used in immune and inflammatory disease research
Catalog No. 50-003-99163 Supplier Medchemexpress LLC Supplier No. HYP583410MG
Boc-AEVD-CHO is a Caspase 8 inhibitor useful in the study of apoptosis and immune and inflammatory diseases.
  • Inhibits Caspase 8
  • Useful in apoptosis studies
  • Used in immune and inflammatory disease research