Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-AEVD-CHO | 220094-15-5 | MFCD00210086 | 98.3% | 516.54 | 5 MG
SDP

Catalog No. 5000399845
Encompass_Preferred

Boc-AEVD-CHO is a Caspase 8 inhibitor useful in the study of apoptosis and immune and inflammatory diseases. This compound is a white to off-white solid with a purity of 98.34%, intended for research use only.

  • Caspase 8 inhibitor
  • Useful for research into apoptosis
  • Useful for research into immune and inflammatory diseases
  • White to off-white solid appearance
  • High purity of 98.34%
  • Molecular weight of 516.54
  • Sequence: Boc-Ala-Glu-Val-Asp-CHO
  • Good solubility in DMSO (66.67 mg/mL)
  • Stable storage conditions for powder and solvent forms

Catalog No. 50-003-99845 Supplier Medchemexpress LLC Supplier No. HYP58345MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-AEVD-CHO is a Caspase 8 inhibitor useful in the study of apoptosis and immune and inflammatory diseases. This compound is a white to off-white solid with a purity of 98.34%, intended for research use only.

  • Caspase 8 inhibitor
  • Useful for research into apoptosis
  • Useful for research into immune and inflammatory diseases
  • White to off-white solid appearance
  • High purity of 98.34%
  • Molecular weight of 516.54
  • Sequence: Boc-Ala-Glu-Val-Asp-CHO
  • Good solubility in DMSO (66.67 mg/mL)
  • Stable storage conditions for powder and solvent forms

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.