Learn More
CPC Scientific Boc-Ala-Glu-Val-Asp-CHO 0.5MG
SDP
Supplier:  CPC Scientific CASP079A
Encompass
Group III caspase inhibitors. This peptide inhibits Caspase-8 with a Ki value of 1.6nM. ONE-LETTER SEQUENCE: Boc-AEVD-CHOMOLECULAR FORMULA: C22H36N4O10MOLECULAR WEIGHT:516.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [220094-15-5], RESEARCH AREA: ApoptosisREFERENCES: M. Garcia-Calvo et al., J. Biol. Chem., 273, 32608 (1998)
Catalog No. 50-210-9068