Learn More
Medchemexpress LLC Boc-β-Ala-ol (3-(Boc-amino)-1-propanol) | 58885-58-8 | MFCD00191883 | 99.8% | 175.23 | 100 G
SDP
Catalog No. 5000404311
Encompass_Preferred
Boc-β-Ala-ol (3-(Boc-amino)-1-propanol) is an alanine derivative featuring a Boc protecting group at the N-terminus. It is utilized in the synthesis of bioactive peptide mimics, such as Nα-Benzoyl-α-azaornithine phenyl ester, which exhibits trypsin inhibitory activity. This product is for research use only.
  • Alanine derivative with Boc protecting group
  • Used in the synthesis of bioactive peptide mimics
  • Exhibits trypsin inhibitory activity
  • Available in various sizes
  • Data sheet, COA, SDS, and handling instructions available
Catalog No. 50-004-04311 Supplier Medchemexpress LLC Supplier No. HY34738100G
Boc-β-Ala-ol (3-(Boc-amino)-1-propanol) is an alanine derivative featuring a Boc protecting group at the N-terminus. It is utilized in the synthesis of bioactive peptide mimics, such as Nα-Benzoyl-α-azaornithine phenyl ester, which exhibits trypsin inhibitory activity. This product is for research use only.
  • Alanine derivative with Boc protecting group
  • Used in the synthesis of bioactive peptide mimics
  • Exhibits trypsin inhibitory activity
  • Available in various sizes
  • Data sheet, COA, SDS, and handling instructions available