Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-amido-PEG9-amine | 890091-43-7 | ≥98.0% | C25H52N2O11 | 1 G
SDP

Catalog No. 5000353012
Encompass_Preferred

Boc-amido-PEG9-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands linked together: one targets an E3 ubiquitin ligase, and the other targets a specific protein. This mechanism leverages the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • PEG-based PROTAC linker
  • Liquid appearance
  • Colorless to light yellow liquid
  • Purity greater than or equal to 98.0%
  • Ships at room temperature in continental US
  • Store at 4°C, protected from light
  • For long-term storage, store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light.

Catalog No. 50-003-53012 Supplier Medchemexpress LLC Supplier No. HY1402331G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-amido-PEG9-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands linked together: one targets an E3 ubiquitin ligase, and the other targets a specific protein. This mechanism leverages the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • PEG-based PROTAC linker
  • Liquid appearance
  • Colorless to light yellow liquid
  • Purity greater than or equal to 98.0%
  • Ships at room temperature in continental US
  • Store at 4°C, protected from light
  • For long-term storage, store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.