Learn More
Medchemexpress LLC Boc-amido-PEG9-amine | 890091-43-7 | ≥98.0% | C25H52N2O11 | 100 MG
SDP
Catalog No. 5000357113
Encompass_Preferred
Boc-amido-PEG9-amine is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are molecules that utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins, by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-57113 Supplier Medchemexpress LLC Supplier No. HY140233100MG
Boc-amido-PEG9-amine is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are molecules that utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins, by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only