Learn More
Medchemexpress LLC Boc-amido-PEG9-amine | 890091-43-7 | ≥98.0% | C25H52N2O11 | 50 MG
SDP
Catalog No. 5000357218
Encompass_Preferred
Boc-amido-PEG9-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed with two distinct ligands connected by a linker, one binding to an E3 ubiquitin ligase and the other to a target protein. This mechanism exploits the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • Can be used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • Ligand for an E3 ubiquitin ligase
  • Ligand for the target protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-57218 Supplier Medchemexpress LLC Supplier No. HY14023350MG
Boc-amido-PEG9-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed with two distinct ligands connected by a linker, one binding to an E3 ubiquitin ligase and the other to a target protein. This mechanism exploits the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • Can be used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • Ligand for an E3 ubiquitin ligase
  • Ligand for the target protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins