Learn More
Medchemexpress LLC (Boc-aminooxy)acetic acid | 42989-85-5 | MFCD01632027 | 98.0% | 191.18 | C7H13NO5 | 10 G
SDP
Catalog No. 5000301472
Encompass_Preferred
(Boc-aminooxy)acetic acid is a Boc-protected aminooxy acetic acid used as a biochemical reagent for bioconjugation and organic synthesis. The aminooxy group enables chemoselective oxime ligation to aldehydes and ketones, while the Boc protecting group allows controlled deprotection and orthogonal coupling strategies. Available in solid and solution formats, the reagent is supplied with quality and safety documentation for laboratory use.
  • Enables chemoselective oxime ligation with aldehydes and ketones.
  • Boc-protected aminooxy functionality for controlled deprotection and coupling.
  • High purity (≥98.0%) suitable for synthesis and labeling.
  • Available as solid and solution formats for flexible handling.
  • Supplied with COA and SDS to support quality and safety compliance.
  • Stable when stored under recommended conditions to preserve activity.
Catalog No. 50-003-01472 Supplier Medchemexpress LLC Supplier No. HYW01962310G
(Boc-aminooxy)acetic acid is a Boc-protected aminooxy acetic acid used as a biochemical reagent for bioconjugation and organic synthesis. The aminooxy group enables chemoselective oxime ligation to aldehydes and ketones, while the Boc protecting group allows controlled deprotection and orthogonal coupling strategies. Available in solid and solution formats, the reagent is supplied with quality and safety documentation for laboratory use.
  • Enables chemoselective oxime ligation with aldehydes and ketones.
  • Boc-protected aminooxy functionality for controlled deprotection and coupling.
  • High purity (≥98.0%) suitable for synthesis and labeling.
  • Available as solid and solution formats for flexible handling.
  • Supplied with COA and SDS to support quality and safety compliance.
  • Stable when stored under recommended conditions to preserve activity.