Learn More
Medchemexpress LLC Boc-aminooxy acetic acid | 42989-85-5 | MFCD01632027 | ≥98.0% | 191.18 g·mol⁻1 | C7H13NO5 | 5 G
SDP
Catalog No. 5000300917
Encompass_Preferred
Boc-aminooxy acetic acid is a Boc-protected aminooxy acetic acid used as a reagent for organic synthesis and bioconjugation. The Boc group protects the aminooxy functionality during synthetic steps; after deprotection the aminooxy group enables oxime ligation to carbonyl-containing biomolecules. It is used for linker and probe synthesis, labeling, and conjugate preparation in research settings.
Catalog No. 50-003-00917 Supplier Medchemexpress LLC Supplier No. HYW0196235G
Boc-aminooxy acetic acid is a Boc-protected aminooxy acetic acid used as a reagent for organic synthesis and bioconjugation. The Boc group protects the aminooxy functionality during synthetic steps; after deprotection the aminooxy group enables oxime ligation to carbonyl-containing biomolecules. It is used for linker and probe synthesis, labeling, and conjugate preparation in research settings.