Learn More
Medchemexpress LLC (Boc-aminooxy)acetic acid | 42989-85-5 | MFCD01632027 | 98.0% | 191.18 g/mol | C7H13NO5 | 1 ML
SDP
Catalog No. 5000301679
Encompass_Preferred
(Boc-aminooxy)acetic acid is a Boc-protected aminooxy acetic acid reagent used to introduce an aminooxy (hydroxylamine) functional group for oxime ligation and bioconjugation. Supplied as a solid and as a 10 mM solution in DMSO, it has CAS 42989-85-5, molecular formula C7H13NO5, molecular weight 191.18 g/mol, and typical purity of 98.0%.
  • Facilitates oxime ligation between aminooxy and aldehyde or ketone groups.
  • Protected Boc group enables orthogonal deprotection strategies.
  • Available as a 10 mM solution in DMSO for immediate use in assays.
  • High purity suitable for research and synthetic applications.
  • Storage-stable when handled under recommended conditions to minimize degradation.
Catalog No. 50-003-01679 Supplier Medchemexpress LLC Supplier No. HYW01962310MM/1ML
(Boc-aminooxy)acetic acid is a Boc-protected aminooxy acetic acid reagent used to introduce an aminooxy (hydroxylamine) functional group for oxime ligation and bioconjugation. Supplied as a solid and as a 10 mM solution in DMSO, it has CAS 42989-85-5, molecular formula C7H13NO5, molecular weight 191.18 g/mol, and typical purity of 98.0%.
  • Facilitates oxime ligation between aminooxy and aldehyde or ketone groups.
  • Protected Boc group enables orthogonal deprotection strategies.
  • Available as a 10 mM solution in DMSO for immediate use in assays.
  • High purity suitable for research and synthetic applications.
  • Storage-stable when handled under recommended conditions to minimize degradation.