Learn More
Medchemexpress LLC Boc-aminooxy-amide-PEG4-propargyl | 2253965-01-2 | 404.46 g/mol | C18H32N2O8 | 100 MG
SDP
Catalog No. 5000203693
Encompass_Preferred
Boc-aminooxy-amide-PEG4-propargyl is a Boc-protected aminooxy-containing PEG4 linker with a propargyl (alkyne) terminus. It is described as a non-cleavable, 4-unit PEG ADC linker used in conjugation and click chemistry applications.
  • Non-cleavable 4-unit PEG linker.
  • Contains Boc-protected aminooxy and propargyl (alkyne) functional groups.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) click reactions.
  • Suitable for antibody-drug conjugate (ADC) synthesis and general bioconjugation.
  • Available in small research pack sizes for synthetic applications.
Catalog No. 50-002-03693 Supplier Medchemexpress LLC Supplier No. HY133436100MG
Boc-aminooxy-amide-PEG4-propargyl is a Boc-protected aminooxy-containing PEG4 linker with a propargyl (alkyne) terminus. It is described as a non-cleavable, 4-unit PEG ADC linker used in conjugation and click chemistry applications.
  • Non-cleavable 4-unit PEG linker.
  • Contains Boc-protected aminooxy and propargyl (alkyne) functional groups.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) click reactions.
  • Suitable for antibody-drug conjugate (ADC) synthesis and general bioconjugation.
  • Available in small research pack sizes for synthetic applications.