Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-Aminooxy-PEG2 | 1807503-86-1 | 97.0% | C9H19NO5 | 100 MG
SDP

Catalog No. 5000357022
Encompass_Preferred

Boc-Aminooxy-PEG2 is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one for an E3 ubiquitin ligase and the other for the target protein, exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Exploits the intracellular ubiquitin-proteasome system for target protein degradation.
  • Molecular weight: 221.25.
  • Molecular formula: C9H19NO5.
  • Appearance: Viscous liquid.
  • Color: Colorless to light yellow.
  • Purity: 97.0%.

Catalog No. 50-003-57022 Supplier Medchemexpress LLC Supplier No. HY140421100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-Aminooxy-PEG2 is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one for an E3 ubiquitin ligase and the other for the target protein, exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Exploits the intracellular ubiquitin-proteasome system for target protein degradation.
  • Molecular weight: 221.25.
  • Molecular formula: C9H19NO5.
  • Appearance: Viscous liquid.
  • Color: Colorless to light yellow.
  • Purity: 97.0%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.