Learn More
Medchemexpress LLC Boc-Aminooxy-PEG2 | 1807503-86-1 | C9H19NO5 | 250 MG
SDP
Catalog No. 5000357233
Encompass_Preferred
Boc-Aminooxy-PEG2 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PROTACs utilize two different ligands connected by a linker.
  • One ligand targets an E3 ubiquitin ligase.
  • The other targets the protein of interest.
Catalog No. 50-003-57233 Supplier Medchemexpress LLC Supplier No. 250MGHY140421
Boc-Aminooxy-PEG2 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PROTACs utilize two different ligands connected by a linker.
  • One ligand targets an E3 ubiquitin ligase.
  • The other targets the protein of interest.