Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-Aminooxy-PEG2 | 1807503-86-1 | MFCD28122959 | 97.0% | C9H19O5N1 | 1 G
SDP

Catalog No. 5000356643
Encompass_Preferred

Boc-Aminooxy-PEG2 is a chemical compound with the CAS Number 1807503-86-1 and MFCD Number MFCD28122959. It has a chemical formula of C9H19O5N1 and a purity of 97.0%. This compound is suitable for various laboratory applications requiring a high-purity reagent. It is recommended to store the pure form at -20°C for 3 years or 4°C for 2 years. When in solvent, it should be stored at -80°C for 6 months or -20°C for 1 month.

  • Chemical formula: C9H19O5N1
  • Purity: 97.0%
  • Pure form storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-56643 Supplier Medchemexpress LLC Supplier No. HY1404211G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-Aminooxy-PEG2 is a chemical compound with the CAS Number 1807503-86-1 and MFCD Number MFCD28122959. It has a chemical formula of C9H19O5N1 and a purity of 97.0%. This compound is suitable for various laboratory applications requiring a high-purity reagent. It is recommended to store the pure form at -20°C for 3 years or 4°C for 2 years. When in solvent, it should be stored at -80°C for 6 months or -20°C for 1 month.

  • Chemical formula: C9H19O5N1
  • Purity: 97.0%
  • Pure form storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.