Learn More
Medchemexpress LLC Boc-Aminooxy-PEG2-C2-amine | 252378-69-1 | 98.0% | C11H24N2O5 | 250 MG
SDP
Catalog No. 5000352771
Encompass_Preferred
Boc-Aminooxy-PEG2-C2-amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is for research use only.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • For research use only
Catalog No. 50-003-52771 Supplier Medchemexpress LLC Supplier No. 250MGHY140426
Boc-Aminooxy-PEG2-C2-amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is for research use only.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • For research use only