Learn More
Medchemexpress LLC Boc-Aminooxy-PEG4-NH2 | 2062663-66-3 | 95.0% | C13H28N2O7 | 100 MG
SDP
Catalog No. 5000356895
Encompass_Preferred
Boc-Aminooxy-PEG4-NH2 is a PEG-based PROTAC linker used in the synthesis of PROTACs. These molecules exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Utilizes intracellular ubiquitin-proteasome system
Catalog No. 50-003-56895 Supplier Medchemexpress LLC Supplier No. HY140438100MG
Boc-Aminooxy-PEG4-NH2 is a PEG-based PROTAC linker used in the synthesis of PROTACs. These molecules exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Utilizes intracellular ubiquitin-proteasome system