Learn More
Medchemexpress LLC Boc-Aminooxy-PEG4-NH2 | 2062663-66-3 | 95.0% | C13H28N2O7 | 50 MG
SDP
Catalog No. 5000353145
Encompass_Preferred
Boc-Aminooxy-PEG4-NH2 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates the degradation of target proteins
Catalog No. 50-003-53145 Supplier Medchemexpress LLC Supplier No. HY14043850MG
Boc-Aminooxy-PEG4-NH2 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates the degradation of target proteins