Learn More
Medchemexpress LLC Boc-aminoxy-PEG4-acid | 2062663-68-5 | 98.0% | C16H31NO9 | 100 MG
SDP
Catalog No. 5000353060
Encompass_Preferred
Boc-aminoxy-PEG4-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs, which are designed to selectively degrade target proteins by leveraging the intracellular ubiquitin-proteasome system. It is intended for research use only.
  • Used in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-53060 Supplier Medchemexpress LLC Supplier No. HY140413100MG
Boc-aminoxy-PEG4-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs, which are designed to selectively degrade target proteins by leveraging the intracellular ubiquitin-proteasome system. It is intended for research use only.
  • Used in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins