Learn More
Medchemexpress LLC Boc-aminoxy-PEG4-acid | 2062663-68-5 | 98.0% | C16H31NO9 | 25 MG
SDP
Catalog No. 5000352613
Encompass_Preferred
Boc-aminoxy-PEG4-acid is a PEG-based PROTAC linker. It can be used in the synthesis of PROTACs, which are compounds containing two different ligands connected by a linker. One ligand is for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Functions as a PEG-based PROTAC linker
  • Useful in the synthesis of PROTAC compounds
  • Facilitates targeted protein degradation via the ubiquitin-proteasome system
  • Connects ligands for E3 ubiquitin ligase and target proteins
Catalog No. 50-003-52613 Supplier Medchemexpress LLC Supplier No. 25MGHY140413
Boc-aminoxy-PEG4-acid is a PEG-based PROTAC linker. It can be used in the synthesis of PROTACs, which are compounds containing two different ligands connected by a linker. One ligand is for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Functions as a PEG-based PROTAC linker
  • Useful in the synthesis of PROTAC compounds
  • Facilitates targeted protein degradation via the ubiquitin-proteasome system
  • Connects ligands for E3 ubiquitin ligase and target proteins