Learn More
Medchemexpress LLC Boc-aminoxy-PEG4-acid | 2062663-68-5 | 98.0% | C16H31NO9 | 50 MG
SDP
Catalog No. 5000356984
Encompass_Preferred
Boc-aminoxy-PEG4-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are bifunctional molecules comprising two distinct ligands connected by a linker: one ligand targets an E3 ubiquitin ligase, and the other binds to a specific target protein. This design enables PROTACs to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins via the ubiquitin-proteasome system
Catalog No. 50-003-56984 Supplier Medchemexpress LLC Supplier No. HY14041350MG
Boc-aminoxy-PEG4-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are bifunctional molecules comprising two distinct ligands connected by a linker: one ligand targets an E3 ubiquitin ligase, and the other binds to a specific target protein. This design enables PROTACs to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins via the ubiquitin-proteasome system