Learn More
Cayman Chemical Boc-Arg(Boc)2-OH, Purity: ≥98%, CAS: 97745-69-2, 500 mg
SDP
Catalog No. 502990824
Encompass_Preferred
An amino acid building block; has been used in the synthesis of amino acid prodrug forms of cytotoxic anthraquinones with anticancer activity
Catalog No. 50-299-0824 Supplier Cayman Chemical Supplier No. 30522500 MG
An amino acid building block; has been used in the synthesis of amino acid prodrug forms of cytotoxic anthraquinones with anticancer activity