Learn More
Medchemexpress LLC Boc-Arg(di-Z)-OH | 51219-19-3 | MFCD00236824 | ≥98.0% | 542.58 | 500 MG
SDP
Catalog No. 5000454579
Encompass_Preferred
Boc-Arg(di-Z)-OH can be used for the synthesis of amino acids and for the research of inhibitors for processing proteinases. It is coupled via the mixed anhydride (MA) with HGlu(OBzl)-Lys(Z)-Arg(Z,Z)-CH2Cl.
  • Synthesis of amino acids
  • Research of inhibitors for processing proteinases
Catalog No. 50-004-54579 Supplier Medchemexpress LLC Supplier No. HYW013097500MG
Boc-Arg(di-Z)-OH can be used for the synthesis of amino acids and for the research of inhibitors for processing proteinases. It is coupled via the mixed anhydride (MA) with HGlu(OBzl)-Lys(Z)-Arg(Z,Z)-CH2Cl.
  • Synthesis of amino acids
  • Research of inhibitors for processing proteinases