Learn More
Medchemexpress LLC Boc-Arg(NO2)-OH | 2188-18-3 | MFCD00065556 | 98.0% | 319.31 | C11H21N5O6 | 100 G
SDP
Catalog No. 5000299788
Encompass_Preferred
Boc-Arg(NO2)-OH is a Boc-protected nitro-arginine derivative used as a building block in peptide synthesis and medicinal chemistry research. Supplied as a solid with high reported purity, it is used as an intermediate for incorporation of nitro-arginine residues into peptides and for chemical modification studies.
  • High purity suitable for research applications.
  • Boc-protected alpha-amino group compatible with standard peptide coupling.
  • Nitro group on the guanidino side chain for activity modulation or selective reduction.
  • Solid powder form for convenient handling and storage.
  • Commonly used as a peptide synthesis building block and chemical intermediate.
Catalog No. 50-002-99788 Supplier Medchemexpress LLC Supplier No. HYW040067100G
Boc-Arg(NO2)-OH is a Boc-protected nitro-arginine derivative used as a building block in peptide synthesis and medicinal chemistry research. Supplied as a solid with high reported purity, it is used as an intermediate for incorporation of nitro-arginine residues into peptides and for chemical modification studies.
  • High purity suitable for research applications.
  • Boc-protected alpha-amino group compatible with standard peptide coupling.
  • Nitro group on the guanidino side chain for activity modulation or selective reduction.
  • Solid powder form for convenient handling and storage.
  • Commonly used as a peptide synthesis building block and chemical intermediate.