Learn More
Carbosynth LLC. Boc-Arg-Val-Arg-Arg-MCA (5 mg vial), 1ea, MW: 842.99, Molecular formula: C38H62N14O8, CAS# 139979-73-0
SDP
Supplier:  Carbosynth LLC. MRR3155V5MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: t-Butyloxycarbonyl-L-Arginyl-L-Valyl-L-Arginyl-L-Arginine 4-Methyl-Coumaryl-7-Amide, Boc-Arg-Val-Arg-Arg-AMC, Boc-RVRR-AMC, Boc-RVRR-MCA, Purity: , Grade A, Application: Substrate for Furin, Storage Temperature: -20C, References: K. Hatsuzawa, K. Murakami, and K. Nakayama, J. Biochem., 111, 296 (1992)., K. Hatsuzawa, M. Nagahara, S. Takahashi, K. Takada, K. Murakami, and K. Nakayama, J. Biol. Chem., 267, 16094, (1992).
Catalog No. 50-168-6021