Learn More
Bachem Boc-Asp(OBzl)-Pro-Arg-AMC · HCl
SDP
Catalog No. 50260324
Encompass_Preferred
Boc-Asp(OBzl)-Pro-Arg-AMC · HCl, 113866-00-5, C37H47N7O9 · HCl, Mr 770.28, 50mg. Highly sensitive substrate for thrombin and trypsin. Kinetic data for Thrombin: kcat = 160 s-1, Km = 11 uM.
Catalog No. 50-260-324 Supplier Bachem Supplier No. 4017304.0050
Boc-Asp(OBzl)-Pro-Arg-AMC · HCl, 113866-00-5, C37H47N7O9 · HCl, Mr 770.28, 50mg. Highly sensitive substrate for thrombin and trypsin. Kinetic data for Thrombin: kcat = 160 s-1, Km = 11 uM.