Learn More
Medchemexpress LLC Boc-Asp(OBzl)-Pro-Arg-AMC (acetate) | 99.2% | 793.86 | 10 MG
SDP
Catalog No. 5000396651
Encompass_Preferred
Boc-Asp(OBzl)-Pro-Arg-AMC acetate functions as a fluorogenic substrate for determining protease activity. This compound undergoes hydrolysis, which then releases 7-amino-4-methylcoumarin (AMC), a fluorescent product that emits a signal under UV light.
  • Fluorogenic substrate for protease activity determination
  • Releases 7-amino-4-methylcoumarin (AMC) upon hydrolysis
  • AMC is fluorescent under UV light
  • Emits a fluorescent signal
Catalog No. 50-003-96651 Supplier Medchemexpress LLC Supplier No. HYP4419A10MG
Boc-Asp(OBzl)-Pro-Arg-AMC acetate functions as a fluorogenic substrate for determining protease activity. This compound undergoes hydrolysis, which then releases 7-amino-4-methylcoumarin (AMC), a fluorescent product that emits a signal under UV light.
  • Fluorogenic substrate for protease activity determination
  • Releases 7-amino-4-methylcoumarin (AMC) upon hydrolysis
  • AMC is fluorescent under UV light
  • Emits a fluorescent signal