Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-Asp(OMe)-fluoromethyl ketone | 187389-53-3 | MFCD03453073 | 98.0% | 263.26 g/mol | C11H18FNO5 | 10 MG
SDP

Catalog No. 5000244763
Encompass_Preferred

Boc-Asp(OMe)-fluoromethyl ketone is a broad-range caspase inhibitor used in vitro to block Fas-mediated phagocytosis and oxidative rupture while sparing IL-8 chemotactic activity. The compound is supplied in multiple sizes and solution formats for research applications.

  • Broad-range caspase inhibitor used for apoptosis and phagocytosis studies.
  • CAS number 187389-53-3 for chemical identification.
  • Molecular weight 263.26 g/mol and formula C11H18FNO5.
  • Purity 98.0% and colorless to light yellow appearance.
  • Storage: sealed, away from moisture; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Available in multiple solid and solution sizes for in vitro use.

Catalog No. 50-002-44763 Supplier Medchemexpress LLC Supplier No. HY10334810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-Asp(OMe)-fluoromethyl ketone is a broad-range caspase inhibitor used in vitro to block Fas-mediated phagocytosis and oxidative rupture while sparing IL-8 chemotactic activity. The compound is supplied in multiple sizes and solution formats for research applications.

  • Broad-range caspase inhibitor used for apoptosis and phagocytosis studies.
  • CAS number 187389-53-3 for chemical identification.
  • Molecular weight 263.26 g/mol and formula C11H18FNO5.
  • Purity 98.0% and colorless to light yellow appearance.
  • Storage: sealed, away from moisture; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Available in multiple solid and solution sizes for in vitro use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.