Learn More
Medchemexpress LLC Boc-Asp(OMe)-fluoromethyl ketone | 187389-53-3 | MFCD03453073 | 98.0% | 263.26 g·mol⁻¹ | C11H18FNO5 | 100 MG
SDP
Catalog No. 5000244747
Encompass_Preferred
Boc-Asp(OMe)-fluoromethyl ketone is a cell-permeable, broad-spectrum caspase inhibitor used as a biochemical tool to study caspase-dependent apoptosis and related processes. It inhibits caspase activity, including Fas-mediated phagocytosis and oxidative rupture, while not affecting IL-8 chemotaxis. The compound is supplied at high purity for research applications.
  • Broad-spectrum caspase inhibitor for apoptosis research.
  • Cell permeable, enabling intracellular enzyme inhibition.
  • High purity (98.0%) for reproducible results.
  • Liquid, colorless to light yellow appearance.
  • Supplied as a 100 MG package for laboratory use.
  • Store sealed, away from moisture; recommended in-solvent storage: -80°C (up to 6 months) or -20°C (up to 1 month).
Catalog No. 50-002-44747 Supplier Medchemexpress LLC Supplier No. HY103348100MG
Boc-Asp(OMe)-fluoromethyl ketone is a cell-permeable, broad-spectrum caspase inhibitor used as a biochemical tool to study caspase-dependent apoptosis and related processes. It inhibits caspase activity, including Fas-mediated phagocytosis and oxidative rupture, while not affecting IL-8 chemotaxis. The compound is supplied at high purity for research applications.
  • Broad-spectrum caspase inhibitor for apoptosis research.
  • Cell permeable, enabling intracellular enzyme inhibition.
  • High purity (98.0%) for reproducible results.
  • Liquid, colorless to light yellow appearance.
  • Supplied as a 100 MG package for laboratory use.
  • Store sealed, away from moisture; recommended in-solvent storage: -80°C (up to 6 months) or -20°C (up to 1 month).