Learn More
Bachem Boc-Asp-OtBu
SDP
Supplier:  Bachem 4026472.0025
Encompass_Preferred
Boc-Asp-OtBu, 34582-32-6, C13H23NO6, Mr 289.33, 25g. Educt for the synthesis of g-mercaptolysine derivatives.
Catalog No. 50-258-396