Learn More
AdipoGen Boc-AspOMe-FMK
SDP
Catalog No. 502562428
Encompass_Preferred
Chemical. CAS 187389-53-3. Formula C11H18FNO5. MW 263.3. Synthetic. Cell permeable, irreversible, broad-spectrum caspase inhibitor. Inhibits non-caspase cysteine proteases, as well as cathepsins H and L. Can be used for in vitro and in vivo studies. Contains a methyl ester group, which facilitates uptake by cells and subsequently is removed by cytoplasmic esterases, leaving the functional inhibitor. Inhibits the pro-apoptotic effect of TNF-alpha IC50=39µM. Shown to reduce the activation of NF-kB, suppresses the phosphorylation of IkBalpha and inhibits TNF-induced expression of ICAM-1 and VCAM-1. Blocks hepatocyte apoptosis in vivo. Neuroprotective.
Catalog No. 50-256-2428 Supplier AdipoGen Supplier No. AGCP30030M001
Chemical. CAS 187389-53-3. Formula C11H18FNO5. MW 263.3. Synthetic. Cell permeable, irreversible, broad-spectrum caspase inhibitor. Inhibits non-caspase cysteine proteases, as well as cathepsins H and L. Can be used for in vitro and in vivo studies. Contains a methyl ester group, which facilitates uptake by cells and subsequently is removed by cytoplasmic esterases, leaving the functional inhibitor. Inhibits the pro-apoptotic effect of TNF-alpha IC50=39µM. Shown to reduce the activation of NF-kB, suppresses the phosphorylation of IkBalpha and inhibits TNF-induced expression of ICAM-1 and VCAM-1. Blocks hepatocyte apoptosis in vivo. Neuroprotective.