Learn More
Medchemexpress LLC Boc-Bip(4,4')-OH | 147923-08-8 | 99.25% | C20H23NO4 | 1 G
SDP
Catalog No. 5000432998
Encompass_Preferred
Boc-Bip(4,4')-OH is a phenylalanine derivative. This compound is commercially used as an ergogenic supplement, known to influence anabolic hormone secretion, improve fuel supply during exercise, enhance mental performance in stress-related tasks, and help prevent exercise-induced muscle damage. It is recognized as a beneficial ergogenic dietary substance.
  • Derived from phenylalanine
  • Functions as an ergogenic supplement
  • Impacts anabolic hormone secretion
  • Supports fuel supply during exercise
  • Improves mental performance under stress
  • Helps prevent exercise-induced muscle damage
  • Soluble in DMSO at 200 mg/mL (ultrasonic required)
  • Intended for research use only
Catalog No. 50-004-32998 Supplier Medchemexpress LLC Supplier No. HY788431G
Boc-Bip(4,4')-OH is a phenylalanine derivative. This compound is commercially used as an ergogenic supplement, known to influence anabolic hormone secretion, improve fuel supply during exercise, enhance mental performance in stress-related tasks, and help prevent exercise-induced muscle damage. It is recognized as a beneficial ergogenic dietary substance.
  • Derived from phenylalanine
  • Functions as an ergogenic supplement
  • Impacts anabolic hormone secretion
  • Supports fuel supply during exercise
  • Improves mental performance under stress
  • Helps prevent exercise-induced muscle damage
  • Soluble in DMSO at 200 mg/mL (ultrasonic required)
  • Intended for research use only