Learn More
Medchemexpress LLC Boc-Bip(4,4')-OH | 147923-08-8 | 99.25% | C20H23NO4 | 10 G
SDP
Catalog No. 5000434837
Encompass_Preferred
Boc-Bip(4,4')-OH is a phenylalanine derivative, intended for research use only and not for sale to patients. Amino acids and their derivatives are commercially used as ergogenic supplements.
  • Phenylalanine derivative
  • Used as an ergogenic supplement
  • Influences secretion of anabolic hormones
  • Supplies fuel during exercise
  • Improves mental performance during stress-related tasks
  • Prevents exercise-induced muscle damage
Catalog No. 50-004-34837 Supplier Medchemexpress LLC Supplier No. HY7884310G
Boc-Bip(4,4')-OH is a phenylalanine derivative, intended for research use only and not for sale to patients. Amino acids and their derivatives are commercially used as ergogenic supplements.
  • Phenylalanine derivative
  • Used as an ergogenic supplement
  • Influences secretion of anabolic hormones
  • Supplies fuel during exercise
  • Improves mental performance during stress-related tasks
  • Prevents exercise-induced muscle damage