Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-bipiperidine-ethynylbenzoic acid | 2308497-81-4 | 99.19% | 412.52 | 25 MG
SDP

Catalog No. 5000350131
Encompass_Preferred

Boc-bipiperidine-ethynylbenzoic acid is an alkyl/ether-based PROTAC linker that can be used in the synthesis of ARD-61. It also acts as a click chemistry reagent, containing an alkyne group capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing azide groups.

  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of ARD-61
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Appears as a white to off-white solid
  • High purity of 99.19%
  • Molecular weight of 412.52

Catalog No. 50-003-50131 Supplier Medchemexpress LLC Supplier No. 25MGHY139661
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-bipiperidine-ethynylbenzoic acid is an alkyl/ether-based PROTAC linker that can be used in the synthesis of ARD-61. It also acts as a click chemistry reagent, containing an alkyne group capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing azide groups.

  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of ARD-61
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Appears as a white to off-white solid
  • High purity of 99.19%
  • Molecular weight of 412.52

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.